ContactUsLinkComponent
Saccharomyces, Budding Cells Slide
Saccharomyces, Budding Cells Slide
Catalog # 470177-440
Supplier:  Avantor
undefined
Saccharomyces, Budding Cells Slide
Catalog # 470177-440
Supplier:  Avantor
Specifications
  • Description:
    Saccharomyces, Budding Cells Slide
  • Cat. No.:
    470177-440
  • Stain:
    Methylene Blue
  • Supplier no.:
    WARD470177-440
  • Orientation:
    Whole mount
Specifications
About this item
Yeast reproduction
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Cellular reproduction by budding.