ContactUsLinkComponent
RNA Removed (sect) mgp
Catalog # 470182-586
Supplier:  Ward's Science
RNA Removed (sect) mgp
Catalog # 470182-586
Supplier:  Ward's Science
Supplier Number:  7932355
Specifications
  • Description:
    RNA Removed (sect) mgp, Each
  • Cat. No.:
    470182-586
  • Supplier no.:
    7932355
Specifications
About this item
Necturus liver. The enzyme RNAase specifically digests RNA, confirming the existence of RNA in the nucleolus of the cytoplasm. Compare to RNA (sect) mgp.