Order Entry
United States
ContactUsLinkComponent
Right Angle Prism
Right Angle Prism
  470021-846
undefined
Right Angle Prism
  470021-846
 :  PAR050

 

  • Description:
    Right Angle Prism
  • Length:
    2"
  • Width:
    1 3/4"
  • Height:
    1"
  • Cat. No.:
    470021-846
  • Supplier no.:
    PAR050

 

 

A must for your Optics lab

  • Clear acrylic
  • Demonstrates refraction and internal reflection

Teach students about the inner workings of the optical instruments they use and explore basic optical properties of light such as refraction, dispersion, and spectral phenomena.