Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Rhodotorula rubra
- Cat. No.:470179-708
- Packaging:Demo Plate
- Media:Sabouraud dextrose agar
- Supplier no.:WARD470179-708
- Incubation conditions:30 °C
Deuteromycete. Pink yeast. Pink to red pigment on agar growth medium. Incubation temperature 30°C. Sabouraud dextrose/YM agar growth medium.
Canada - Biosafety Level 1.