Rhizopus, Sporangia Slide
Catalog # 470176-852
Supplier: Avantor
Specifications
- Description:Rhizopus, Sporangia Slide
- Cat. No.:470176-852
- Stain:Fast Green and Mallory's
- Supplier no.:WARD470176-852
- Orientation:Whole mount
Specifications
About this item
Fungal reproduction
- Identifying features clearly distinguishable
- Rigorous quality control standards
Black bread mold. Shows sporangia used in asexual reproduction.