Replica Diffraction Gratings
About this item
These simple slide-mounted diffraction gratings are great for experiments with lasers and LEDs.
Mounted in 2×2" slides
- 5360 lines per cm
These replica transmission diffraction gratings are transparent and irridescent. Approximately 5400 lines per centimeter, they are perfect for Young's double slit experiment or for viewing gas emission spectra. Mounted in unmarked cardboard slide holders, they are inexpensive and durable.
1 Options Available Below
Product Details & Documents
These simple slide-mounted diffraction gratings are great for experiments with lasers and LEDs.
Mounted in 2×2" slides
- 5360 lines per cm
These replica transmission diffraction gratings are transparent and irridescent. Approximately 5400 lines per centimeter, they are perfect for Young's double slit experiment or for viewing gas emission spectra. Mounted in unmarked cardboard slide holders, they are inexpensive and durable.
Product Family Options
Product Information
- AvailabilityPrice
- 470006-576Diffraction grating slide, 5400 lines/cm, pack of 25Diffraction grating slide, 5400 lines/cm, pack of 25Discontinued-Supplier Number: 1503-00/25
- 470006-578Diffraction grating slide, 5400 lines/cm, eachSupplier Number: 1503-00