Orders LinkContactUsLinkComponent
Radiation Lab Activity
Radiation Lab Activity
Supplier:   The Science Source
Radiation Lab Activity
Supplier:  The Science Source
Certificates
Radiation Lab Activity
1 Options Available Below
Product Details & Documents
Students investigate the absorption and radiation of energy on different surfaces.
  • Activity Includes Light Source
  • Two Experiments in One Activity
  • Students Research Absorption and Radiation of Energy
They then turn on the light at 10 cm distance and observe how temperatures respond differently in the black can and silver can.
In the reverse experiment, the rates of cooling can be determined after students turn off the light.
Ordering information: Each kit includes a 16 oz. black metal and a 16 oz. shiny metal can, two insulating lids, and two other thermometers (-20° to 50°C). The single kit also includes one 120VAC clamp lamp with a reflector and 100 watt light bulb; the class kit includes four lamps.
Product Family Options
Activity
Product Information
  • No. of lab groups
    Availability
    Price
  • 470002-500
    Radiation Lab Activity, Materials for Eight Lab Groups
    Radiation Lab Activity, Materials for Eight Lab Groups
    8
    Discontinued
    Supplier Number: -
  • 470006-730
    Radiation Lab Activity, Materials for One Lab Group
    Radiation Lab Activity, Materials for One Lab Group
    1
    Discontinued
    Supplier Number: -
  • 470201-200
    Lab Activity, Radiation Kit, ESCP, For 15 Student Groups
    Lab Activity, Radiation Kit, ESCP, For 15 Student Groups
    15
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $661.00
    $661.00
    Kit of 1
       $661.00
    Supplier Number: 16226
Lamp for Activity
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Clamp lamp with reflector
    Log in for availability
    Log in for availability
    Each
    Each - $19.45
    $19.45
    Log in for availability
    Case of 12
    Case of 12 - $19.45
    $19.45
    Each
       $19.45
    Case of 12
       $19.45
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Clamp lamp with reflector
    Clamp lamp with reflector
    470004-224
    Log in for availability
    Each - $19.45