Specifications
- Description:Qualitative ELISA Kit
- Cat. No.:470120-906
- Supplier no.:278-B
Specifications
About this item
Demonstrate the quantitation of varying concentrations of viral antigens as detected by the intensity of the color reaction due to the accumulation of products.
- Material for Six Groups with 12 Reactions Each
- Instructions with Background Information Included
- Some Components Require 4°C Refrigeration
The kit provides material for six groups with 12 reactions each.
Some components require 4°C refrigeration upon receipt.