ContactUsLinkComponent
Ward's® Protozoa Classification Set
Ward's® Protozoa Classification Set
Catalog # 470180-224
Supplier:  Avantor
undefined
Ward's® Protozoa Classification Set
Catalog # 470180-224
Supplier:  Avantor
Specifications
  • Description:
    WARD’S Protozoa Classification Set
  • Cat. No.:
    470180-224
  • Supplier no.:
    WARD470180-224
Specifications
About this item
One Culture from Each of the Four Major Types
Study a variety of animal-like protists that lack chlorophyll and ingest food as animals do. The set of four includes cultures of Amoeba proteus (amoeba), Chilomonas (zooflagellate), Gregarines in meal-worm host (sporozoan), and Paramecium (ciliate), as well as the Working with Protozoa manual and materials for 35 students.