Protist Structure Kit for 30 Students
Catalog # 470176-504
Supplier: Avantor
Specifications
- Description:Protist Structure Kit for 30 Students
- Cat. No.:470176-504
- Supplier no.:WARD470176-504
Specifications
About this item
Contains Collections of Specimens Used Most Often in Biology Classes
- Study various types of protists
- Live specimen
- Materials for 30 students
Protist Structure Kit is made up of diverse specimens representing various types of protists. Includes live Paramecium, Amoeba and Euglena, study guides for each organism, and structure of protozoa literature. Also includes protist slowing agent, vital stains (Methylene Blue and Janus Green) and pipets. Contains enough materials for up to 30 students.