About this item
Isolated from human urinary tract and wound infections, this organism is identifiable by its characteristic putrefactive odor.
- Biosafety Level: 2
- Canada: Containment Level 2 - PHAC Licence required
- Motility: Positive
- Morphology: Rods
- Gram Reaction: Negative
- Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout