Ward's® Preying Protist Study Set
Catalog # 470180-204
Supplier: Avantor
Specifications
- Description:WARD’S Preying Protist Study Set
- Cat. No.:470180-204
- Supplier no.:WARD470180-204
Specifications
About this item
Observe Two Protists Hunting Included Prey
- Study preying protists
- Includes two types
- Manual included
Simply add the Paramecium provided and watch the Chaos and Didinium specimens stalk, capture, and assimilate them. The set of three includes Chaos, Didinium, and Paramecium (prey).
**Please see the product resources tab for "Working with Protozoa" manual.