Vernier® Power Amplifier Accessory Speaker
470017-522
- Description:Power Amplifier Accessory Speaker
- Cat. No.:470017-522
- Supplier no.:PAAS-PAMP
This kit includes a speaker and accessories used with the Vernier Power Amplifier to study mechanical waves and springs. The speaker contains a lightweight metal disc glued to the speaker cone. A drive post attached to the metal plate also allows you to drive elastic materials such as strings and springs.