Portable Lab Light
Lab light
Catalog # 470144-954
Supplier: O. C. WHITE CO.
Supplier #: 12400
Jump to:
Product Details & Documents
Adjustable lighting for lab settings. The unique domed, vented shade provides cool, direct 60 W light.
- Arm extends to 8" vertically and 12" horizontally
- Arm rotates 360°
- Features a felt-bottomed base for stability
Bulb not included.
Specifications
- Description:Lab light
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



