Polytrichum
Catalog # 470176-476
Supplier: NORTHEAST BIOLOGICAL PROCESSOR
Specifications
- Description:Polytrichum
- Cat. No.:470176-476
- Supplier no.:POLY3-4
Specifications
About this item
Polytrichum sp. Haircap moss. A widely studied example of a typical moss, with leafy, upright growth habit that shows alternation of generations. Appearance may vary, depending on season; it may show antheridia, archegonia, or sporophyte. It is supplied in a plastic bag with care instructions. Size: 3" x 4".