Order Entry
United States
ContactUsLinkComponent
Polymethylpentene Petri Dish
Polymethylpentene Petri Dish
Catalog # 470211-858
undefined
Polymethylpentene Petri Dish
Catalog # 470211-858
Supplier Number:  5500-0010

Specifications

  • Description:
    Polymethylpentene Petri Dish, 100mm x 15mm
  • Environmentally Preferable:
  • Cat. No.:
    470211-858
  • Supplier no.:
    5500-0010

Specifications

About this item

Transparent, shatterproof, and reusable plastic dishes are useful for applications requiring resistance to most acids, bases, alcohols, esters, and aldehydes. Ridges on the top interlock with the bottom edges for stacking. Autoclavable. Size: 100 mm dia. x 15 mm deep.