Plastic Cylinders
Supplier: DEV-COA
About this item
These cylinders are made of Tenite® for moderate chemical resistance.
- Removable Square Base
- Handy Pour Spout
- Tenite and Plastic Construction
- For Use in Pre-High School Science Programs
- Easy-to-Read Single Graduations with Yellow Background
They are for use in pre-high school science programs.
Each cylinder has easy-to-read graduations and a convenient pour spout.
2 Options Available Below
Product Details & Documents
These cylinders are made of Tenite® for moderate chemical resistance.
- Removable Square Base
- Handy Pour Spout
- Tenite and Plastic Construction
- For Use in Pre-High School Science Programs
- Easy-to-Read Single Graduations with Yellow Background
They are for use in pre-high school science programs.
Each cylinder has easy-to-read graduations and a convenient pour spout.
Specifications for Product Family
Specifications
- Catalog No:
- 470121-886
- 470121-888
- 470121-890
- 470121-892
- 470121-894
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
Specifications
Product Family Options
Product Information
- CapacityGraduation IntervalAvailabilityPrice
- Supplier Number: DPS-0010Specifications:
More
Specifications
Cat. No.470121-886FormTall - 25 ml0.5 mLDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.470121-888FormTall - Supplier Number: DPS-0012Specifications:
More
Specifications
Cat. No.470121-890FormTall - 100 ml1 mLDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.470121-892FormTall - 250 ml2 mLDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.470121-894FormTall