- Description:Phycomyces blakesleeanus (-) Tube Culture
- Cat. No.:470179-216
- Supplier no.:WARD470179-216
Zygomycete. Mating strain; cross with Phycomyces blakesleeanus (+) to produce zygospores. Incubation temperature 25°C. Potato dextrose agar growth medium.