Ward's® Philodina
Catalog # 470176-460
Supplier: Ward's Science
Specifications
- Description:Philodina, For 35 Students
- Environmentally Preferable:
- Cat. No.:470176-460
- Supplier no.:WARD470176-460
Specifications
About this item
Philodina sp. Easy to culture and easy to examine under a microscope because of the lack of a lorica (shell), these typical rotifers are perfect for general study. Culture instructions are included.