Ward's® Phacus
Phacus Culture for 35 Students
Catalog # 470179-898
Supplier: Ward's Science
Supplier #: PHAC30/35
Jump to:
Product Details & Documents
Freshwater. Unicellular; heart-shaped cell. Soil-water medium.
Characterized by a lack of rigid cellulosic walls, instead these organisms have pellicles made of protein. They store paramylon instead of starch.
Documents
Free_Living_Protozoa Protists Working_with_Protozoa
Product Literature
Specifications
- Description:Phacus Culture for 35 Students