Order Entry
United States
ContactUsLinkComponent
 
Ward's® Petrographic Microscope Slides
Catalog # 470150-480
Supplier:  Erie Scientific
undefined
Ward's® Petrographic Microscope Slides
Catalog # 470150-480
Supplier:  Erie Scientific
Supplier Number:  2746-602765

Specifications

  • Description:
    Ward’s petrographic slides
  • Environmentally Preferable:
  • Cat. No.:
    470150-480
  • Supplier no.:
    WRD-2746

Specifications

About this item

Manufactured to Precise Specifications

Each noncorrosive glass slide, uniform in thickness, length, and width, measures 1" x 2", 27 x 46 mm.

Edges are beveled, ground, and polished.

Thickness: 1.2 mm.