ContactUsLinkComponent
PCR-Based DNA Fingerprinting Kit
PCR-Based DNA Fingerprinting Kit
Catalog # 470120-896
Supplier:  Edvotek
undefined
PCR-Based DNA Fingerprinting Kit
Catalog # 470120-896
Supplier:  Edvotek
Supplier Number:  371
Specifications
  • Description:
    PCR-Based DNA Fingerprinting Kit
  • Environmentally Preferable:
  • No. of lab groups:
    5
  • Cat. No.:
    470120-896
  • Supplier no.:
    371
Specifications
About this item
Students explore Forensic Science with the PCR-Based DNA Fingerprinting Kit.
  • Lab Encourages Independent Inquiry
  • Time Required: 75 Minutes plus Two Hours for PCR
  • Students Practice PCR in the Classroom
  • Instructions Included
  • PCR EdvoBeads Included
This kit provides easy-to-follow instructions, so students can develop various crime scene scenarios independently.
Plasmid DNA, when amplified by PCR, provides materials that represent individual DNA profiles. Students use these outcomes to solve a crime.
Ordering information: This kit includes instructions, PCR EdvoBeads, DNA templates, primers, DNA ladder, ultrapure water, wax beads, agarose, loading dye, electrophoresis buffer, InstaStain Ethidium Bromide and FlashBlue stain. Micropipettes that measure between 5 and 50 µl, tips, a thermal cycler, electrophoresis apparatus, a power supply, a microwave or hot plate, and a UV transilluminator are required but not included.
Packaging: Some components require freezer storage on arrival.