Pancreas, Mammal Slide
Catalog # 470177-620
Supplier: Avantor
Specifications
- Description:Pancreas, Mammal Slide
- Environmentally Preferable:
- Cat. No.:470177-620
- Stain:Hematoxylin and Eosin
- Supplier no.:WARD470177-620
- Orientation:Section
Specifications
About this item
Digestive organ and endocrine tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Showing secretory acini, ducts, and islets of Langerhans.