ContactUsLinkComponent
Olfactory Epithelium Slide
Olfactory Epithelium Slide
  470182-744
 :  Avantor
undefined
Olfactory Epithelium Slide
  470182-744
 :  Avantor
  • Description:
    Olfactory Epithelium Slide
  • Cat. No.:
    470182-744
  • Stain:
    Hematoxylin and Eosin
  • Supplier no.:
    WARD470182-744
  • Orientation:
    Section
Illustrates specialized nasal tissue
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows pseudostratified columnar epithelium with no goblet cells and no distinct basal lamina.