ContactUsLinkComponent
O₂ Gas Sensor to Spirometer Adapter
O₂ Gas Sensor to Spirometer Adapter
  470133-506
undefined
O₂ Gas Sensor to Spirometer Adapter
  470133-506
 :  O2-SPR
  • Description:
    O2 Gas Sensor to Spirometer
  • Cat. No.:
    470133-506
  • Supplier no.:
    O2-SPR
This adapter facilitates the connection between an Oxygen Gas Sensor and a Spirometer. Measures the oxygen gas concentration and flow rate of exhaled air.