- Description:Nonmetallic minerals, Topic Set
- No. of samples:12
- Cat. No.:470025-362
- Supplier no.:WARD470025-362
Substantial Set of Economically Important Minerals
- Developed by teachers and WARD's Staff Geologists
- Classroom tested for dependabilty
- Designed to save valuble planning and preparation time
Students will learn the economic importance of nonmetallic minerals with this comprehensive GEO-logic set. A teacher's guide and student copymasters are included for the following activities: grouping according to type, uses based on chemical and physical properties, and importance of building minerals. The set is housed in a compartmented collection box.
GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides. The types of GEO-logic teaching collections include:
Topic Sets, which provide greater depth in an individual subject area and include both a teacher’s guide and student copymasters
Contents: 12 numbered samples, 1×2" Minerals
Rocks:apatite, fluorite, garnet, granite, graphite, gypsum, kaolin, limestone, mica, quartz, sulfur, talc