ContactUsLinkComponent
Ward's® New York Vikings: A Lab Activity in Forensic Anthropology and Geology
Ward's® New York Vikings: A Lab Activity in Forensic Anthropology and Geology
Catalog # 470125-674
Supplier:  Avantor
undefined
Ward's® New York Vikings: A Lab Activity in Forensic Anthropology and Geology
Catalog # 470125-674
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    WARD'S New York Vikings: A Lab Activity in Forensic Anthropology and Geology
  • No. of lab groups:
    3
  • Cat. No.:
    470125-674
  • Supplier no.:
    WARD470125-674
Specifications
About this item
Analyze human bone, artifacts, and rocks.
  • Grades 9-12
  • Real World Case Based Scenario
  • Estimated class/lab time required: 180 minutes over 4 days
In this forensic science activity, students will apply hands-on forensic techniques to analyze a scenario involving multiple lines of anthropological, archaeological, and geological evidence from human bone replicas, artifact photos, and rocks. A human skeleton found in the woods in northern New York and associated artifacts and rocks suggest a possible Viking burial. But is it? Or is it a creative cover-up for a murder scene? Three student teams work together to analyze the evidence, reconstruct the scene, and determine if a crime was committed and, if so, the identity of the perpetrator. This detailed and fascinating study of bones and stones was developed in collaboration with Tyler G. O’Brien, Ph. D., Associate Professor of Anthropology at the University of Northern Iowa, and Neal R. O-Brien, Emeritus Professor of Geology at SUNY at Potsdam, for students in high school and early college. The activity can be completed in four 45-minute lab sessions with additional research time outside of class.
Materials include:
Replicas of three adult human bones (skull, femur and pelvis)
Metric vernier caliper
Caliper
Set of six rocks
Limestone
Magnifiers
Pipettes
Hydrochloric acid
Literature
Also needed are a computer with Internet access
Atlas of the human skeleton
World map