Neuroglia, Fibrous Astrocytes Slide
470182-686
: Avantor
- Description:Neuroglia, Fibrous Astrocytes Slide
- Cat. No.:470182-686
- Stain:Silver stain
- Supplier no.:WARD470182-686
- Orientation:Section
Commonly found in white matter
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal cerebrum. Shows cortex treated with Cajal’s gold sublimate.