ContactUsLinkComponent
Motor Nerve Endings in Muscle (Reptile) Slide
Motor Nerve Endings in Muscle (Reptile) Slide
Catalog # 470177-602
Supplier:  Avantor
undefined
Motor Nerve Endings in Muscle (Reptile) Slide
Catalog # 470177-602
Supplier:  Avantor
Specifications
  • Description:
    Motor Nerve Endings in Muscle (Reptile)
  • Cat. No.:
    470177-602
  • Stain:
    Gold chloride and formic acid
  • Supplier no.:
    WARD470177-602
  • Orientation:
    Whole mount
Specifications
About this item
Investigate Nerve Tissue.
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Gold chloride and formic acid stain, whole mount. Showing intercostal muscles and motor nerve plates.