Orders LinkContactUsLinkComponent
G.S.C. International Neon Wand
G.S.C. International Neon Wand
Neon Wand
Catalog # 470201-082
Supplier:   GSC International
Supplier #:  1401
G.S.C. International Neon Wand
Catalog # 470201-082
Supplier:  GSC International
G.S.C. International Neon Wand
G.S.C. International Neon Wand
Neon Wand
Price
$20.00
Log in for availability
Each
Product Details & Documents
Glows with Characteristic Brilliance!
  • Contains neon at reduced pressure.
The neon gas glows when the tube is held in, or passed through a high frequency field, or a high voltage field, even though the tube has no electrodes.
Please Note: Not recommended for use with anything that produces a voltage higher than 75,000 volts.
Specifications
  • Description:
    Neon Wand