Lab-Aids® Nematode Study Kit
Nematode Study Kit, 10
470023-798
: Lab-Aids
: 34
Extract roundworms from soil for up-close study
- Experience and study live, harmless nematodes
- Explore characteristics and structure of nematodes
- Develop microscope technique
- Time required: two 50 minutes class period
This kit offers students a very effective means for learning about living organisms. Students extract nematodes and explore their characteristics and structure. Sampling is easy as Nematodes can be extracted from almost any type of soil sample. Everything needed to extract nematodes is included in this kit. It will require 24 hours to extract nematodes.
: See datasheet for contents information.
- Description:Nematode Study Kit
- No. of lab groups:10