ContactUsLinkComponent
Muscle Types, Composite, Human Slide
Muscle Types, Composite, Human Slide
Catalog # 470177-008
Supplier:  Avantor
undefined
Muscle Types, Composite, Human Slide
Catalog # 470177-008
Supplier:  Avantor
Specifications
  • Description:
    Muscle Types, Composite, Human Slide
  • Environmentally Preferable:
  • Cat. No.:
    470177-008
  • Stain:
    Hematoxylin and Eosin
  • Supplier no.:
    WARD470177-008
  • Orientation:
    Section
Specifications
About this item
Illustrates the differences between muscle types
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Smooth, cardiac, and skeletal muscle.