To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Morganella morganii
- Cat. No.:470179-130
- Packaging:Tube
- Media:Tryptic soy agar
- Supplier no.:WARD470179-130
- Incubation conditions:37 °C
Isolated from human intestine, this organism is known for its distinctive putrefactive odor.
- Biosafety Level: 2
- Canada: Containment Level 2 - PHAC Licence required
- Motility: Positive
- Morphology: Rods
- Gram Reaction: Negative
- Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.