ContactUsLinkComponent
Mohs’ Hardness Collection
Mohs’ Hardness Collection
Catalog # 470016-500
Supplier:  Avantor
undefined
Mohs’ Hardness Collection
Catalog # 470016-500
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Moh's Hardness Kit
  • No. of samples:
    9
  • Cat. No.:
    470016-500
  • Supplier no.:
    WARD470016-500
Specifications
About this item
Unlike many other hardness kits, this kit has only pure crystalline minerals to ensure consistent results when testing.
  • Hand-Picked Samples
  • Housed in a Collection Box
  • Kit Has Only Pure Crystalline Minerals
Additional tools in the kit also allow students to test other physical properties.
The hand-picked samples and housed in a collection box.
Ordering information: This kit includes nine samples (talc through corundum), streak plate, glass plate, copper strip, and a magnet.