ContactUsLinkComponent
Wards® Mohs' Hardness Scale Collection
Wards® Mohs' Hardness Scale Collection
Catalog # 470025-244
Supplier:  Avantor
undefined
Wards® Mohs' Hardness Scale Collection
Catalog # 470025-244
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Mohs' Hardness Scale Collection
  • Cat. No.:
    470025-244
  • Supplier no.:
    WARD470025-244
Specifications
About this item
Our Most Popular collection.
  • 9 numbered samples
  • Includes streak plate, nail, glass slide, and copper strip
All samples are pure crystalline forms to ensure accurate test results. You can also perform other physical properties tests with the included materials. The set is housed in a compartmented collection box, and an identification list is included.