ContactUsLinkComponent
Mini Stirrer
  470019-666
undefined
Mini Stirrer
  470019-666
 :  HI200M-1
  • Description:
    Mini stirrer, stainless steel cover
  • Cat. No.:
    470019-666
  • Electrical:
    110/115 V, 50/60 Hz
  • Top plate dimensions:
    4.75×4.75"
  • Supplier no.:
    HI200MU
Control stirring speed with precision.
  • Both plastic and steel surfaces are chemically resistant
  • Speed limiter for safety with max speed of 1000 rpm
  • 1 L capacity