ContactUsLinkComponent
Ward's® Mineral and Rock Identification Collection
Ward's® Mineral and Rock Identification Collection
Catalog # 470025-154
Supplier:  Avantor
undefined
Ward's® Mineral and Rock Identification Collection
Catalog # 470025-154
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mineral and rock identification collection #2
  • Cat. No.:
    470025-154
  • Supplier no.:
    WARD470025-154
Specifications
About this item
Previously known as the syracuse university mineral and rock set.
  • Housed in a shipping box
  • Identification list included
  • Forty-seven 1×2" unnumbered specimens
  • Streak plate included
Proven successful in a number of introductory college geology courses and high school earth science classes, this set contains high-quality specimens of minerals and rocks, unnumbered, so students can learn to identify them with the help of the instructor.
Minerals: barite, bauxite, biotite, calcite, chalcopyrite, fluorite, galena, garnet, graphite, gypsum (massive), gypsum (selenite), halite, hematite (massive), hermatite (oolitic), hematite (specular), hornblende, labradorite, limonite, magnetite, microcline, muscovite, olivine, pyrite, quartz (rock crystal), rose quartz, sphalerite, sulfur.
Igneous: basalt, diorite, gabbro, granite, granite porphyry, obsidian, peridotite, pumice, syenite.
Sedimentary: conglomerate, limestone, sandstone, shale.
Metamorphic: anthracite, gneiss, hornfels, marble, quartzite, schist, slate.