Specifications
- Description:Mineral and Rock Identification Collection #1
- Cat. No.:470025-184
- Supplier no.:WARD470025-184
Specifications
About this item
Previously Known as Ohio State University Mineral and Rock Set
Recommended for high school earth science classes as well as college-level geology courses.
The specimens are not numbered, so the student, under the guidance of the instructor, can learn how to identify them.
The set is housed in a shipping box, and an identification list is included.Contents: 38 unnumbered samples, 1×1"; streak plateMinerals:biotite, calcite, galena, garnet, halite, hematite, hornblende, limonite, magnetite, milky quartz, muscovite, olivine, orthoclase, plagioclase, pyrite, selenite, serpentine, talcRocks:Igneous: basalt, felsite, gabbro, coarse granite, fine granite, obsidian, pumice, syenite Sedimentary: chert, conglomerate, dolomite, gypsum, limestone, sandstone, shale Metamorphic: gneiss, marble, quartzite, schist, slate