Specifications
- Description:Microbes at Work: Production of an Antibiotic Kit
- No. of lab groups:2
- Cat. No.:470024-104
Specifications
About this item
Explore Fermentation Without Additional Equipment
- Materials for 2 lab groups
- Time Required: ~3hrs. over 5 days
Students gain experience with fermentation, the process by which important medical substances such as antibiotics, interferons, insulin, and growth hormone are produced, by actually growing Bacillus licheniformi in a controlled environment. Students prepare inoculum, measure pH, and test the effectiveness of the antibiotic produced. You will get all of the materials needed to construct a fermenter, enough bacterial culture and media for up to two fermentation runs, teacher’s manual, and student guides. Other cultures can be purchased separately.