Mercury(II) Chloride
Supplier: Ward's Science
Certificates
About this item
CAS Number: 7487-94-7
Formula Weight: 271.50
Formula: HgCl2
Hazard Info: Highly Toxic
Density (g/mL): 5.44
Boiling Point (°C): 300
Freezing Point (°C): 277
Solubility: Water, Alcohol, and Acetic Acid
Synonyms: Mercuric Chloride, Mercury Bichloride, Mercury Dichloride
Shelf Life (months): 12
Storage: Blue
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Frequently Bought Together