ContactUsLinkComponent
Magnetic Field Demonstrator–Iron Rod Type
Magnetic Field Demonstrator–Iron Rod Type
Catalog # 470016-024
Supplier:  Wards
undefined
Magnetic Field Demonstrator–Iron Rod Type
Catalog # 470016-024
Supplier:  Wards
Supplier Number:  PH0802MF2
Specifications
  • Description:
    Magnetic Field Demonstrator–Iron Rod Type
  • Cat. No.:
    470016-024
  • Supplier no.:
    PH0802MF2
Specifications
About this item
A great visual for the classroom
  • Size: 26 x 26 x 1cm
  • The transparency of plates allows for overhead demonstrations
Plates with matrices of 98 small circular chambers contain an iron rod, which functions as effectively as a plotting compass. The transparent plates can be arranged in various ways including as faces of a cube or as a single large two-dimensional surface. This versatility allows you to show the reconfiguration of magnetic fields in one or two dimensions.