Life Science Student Lab Notebook
Life science student lab notebook, 70, 280 mm (11"), 220 mm (8.7")
Catalog # 470023-064
Jump to:
Discontinued
Product Details & Documents
Use this notebook as an additional learning tool to assist students in developing an awareness and discipline for science that mimics that of professional scientists.
- Each notebook has a table of contents for students to log their experiments
- 70 carbonless sets of pages that are sequentially numbered micro-perforated on the bottom copy
The notebook also contains safety information, notebook guidelines, and a variety of life science reference information.
Specifications
- Description:Life science student lab notebook
- Length:220 mm (8.7")
- No. of pages:70
- Width:280 mm (11")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



