Eisco® Large Plastic Dynamics Cart
Catalog # 470039-912
Supplier: Wards
Specifications
- Description:Large plastic dynamics carts, pair
- Cat. No.:470039-912
- Supplier no.:PH0351PL-N8
Specifications
About this item
With the Large Plastic Dynamics Cart, students can explore the changes in motion that occur in a dynamics cart when different amounts of net force are applied.
- Durable Construction
- Mounted on Low-Friction Wheels
- For Elastic and Inelastic-Collision Investigation
- Stackable
- Two Ways to Increase Mass
The mass of each cart can be increased in two ways. The top of each cart is cushioned, enabling the addition of more masses. Users can also stack the carts to make the total mass of the cart two to three-times greater than its basic weight.
Inelastic collisions can also be investigated through the Velcro pads, which are found on the back of each cart. This cart is mounted on a set of three low-friction nylon wheels, which allows it to travel in a straight line over long distances.