ContactUsLinkComponent
LaMotte® Acid Rain Study Kit
LaMotte® Acid Rain Study Kit
  470003-688
 :  LaMotte
undefined
LaMotte® Acid Rain Study Kit
  470003-688
 :  LaMotte
 :  3604-01
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    LaMotte® Acid Rain Study Kit
  • No. of lab groups:
    50
  • Cat. No.:
    470003-688
  • Supplier no.:
    3604-01
Collect and Test Local Rainfall
  • Collect local rainwater samples
  • Measure water acidity
  • Includes a study guide
Your class of environmentalists will be completely equipped to test the rain in your area with this study outfit. They'll use a rain gauge with ground stake holder, pH kit measuring pH 3.0 - 6.5, octet comparator, and reagents. The kit comes packed in a waterproof plastic case with a stimulating study guide and suggested activities. It includes enough materials for 50 tests.
WARNING! This set contains chemicals that may be harmful if misused. Read cautions on individual containers carefully. Not to be used by children except under adult supervision.