Lambda DNA/EcoR I, Hind III Digest
470180-832
:
- Description:Lambda DNA/EcoR I, Hind III Digest, 100μg
- Cat. No.:470180-832
- Supplier no.:G1731
These high-quality DNA Markers were created by digesting specified DNA to completion with one or more restriction enzymes, complete with heat inactivation. The recommended loading for each is 1 {mu lower case}l per lane. Note: Perishable. Store frozen. Shipped next-day air on ice.
Supplied with 1 ml 6X blue/orange loading dye. 100μg (0.5μg/μL).