Kocuria rhizophila (Micrococcus luteus) Slide
Catalog # 470180-874
Supplier: Avantor
Specifications
- Description:Kocuria rhizophila (Micrococcus luteus)
- Cat. No.:470180-874
- Stain:Gram (+)
- Supplier no.:WARD470180-874
- Orientation:Smear
Specifications
About this item
Illustrates an Example of Normal Skin Flora
- Identifying features clearly distinguishable
- Rigorous quality control standards
Spheres in tetrads. It is found on mammalian skin as normal flora.