To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Know Your Minerals and Rocks Collection
- Cat. No.:470025-170
- Supplier no.:WARD470025-170
Specifications
About this item
Everything Students Need to Work Independently to Identify Rocks and Minerals
Step-by-step instructions lead students through the identification of 33 'unknown' mineral and rock samples; they examine physical properties including hardness, cleavage/fracture, tenacity, streak, texture, and grain size, then form their own identification key.
The set is housed in a compartmented collection box, and a comprehensive study guide is included.
Contents:33 unnumbered samples, 1×1"; streak plate; magnifier; numbering setMinerals:
Hardness: apatite, calcite, feldspar, fluorite, gypsum, quartz, talc
Cleavage/Fracture: calcite, feldspar, halite, mica, quartz (obsidian), serpentine
Tenacity: copper, mica, selenite, sulfur
Streak: graphite, hematite, limonite, pyriteRocks:
Igneous: basalt, granite, pumice, rhyolite
Sedimentary: coal, limestone, sandstone, shale
Metamorphic: marble, quartzite, schist, slate.