ContactUsLinkComponent
Introduction to pH Measurement Lab Activity
Introduction to pH Measurement Lab Activity
Catalog # 470150-612
Supplier:  Lab-Aids
undefined
Introduction to pH Measurement Lab Activity
Catalog # 470150-612
Supplier:  Lab-Aids
Supplier Number:  80
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Introduction to pH Measurement Lab Activity
  • Environmentally Preferable:
  • No. of lab groups:
    12
  • Cat. No.:
    470150-612
  • Supplier no.:
    80
Specifications
About this item
Highly Graphic Introduction to the Properties of Acids and Bases
  • Observe the effects of acids and bases on indicators
  • Measure the important function of respiration
  • Requires one 50 minute class period to complete
This series of simple, highly visual lab activities introduce students to the effects of acids and bases on litmus indicators, indicator solutions, and natural indicators. The activities begin by discussing the differences between acids and bases. It then moves to a discussion on the use of pH test paper and the interpretation of their results. Students are also introduced to the concept that acid and base discussions are not limited to chemistry but also extend to applications in biology and the study of the human body. The lab activity uses LAB-AIDS® Chemplates® to study side by side, the reaction of Hydrochloric Acid and Sodium Hydroxide when mixed with other liquids and on pH paper.
Ordering information: Kit includes Chemplates® w/spatula, Hydrochloric acid, Bromothymol blue, Phenol red, Sodium hydroxide, Phenolphthalein, Blue litmus paper, Red litmus paper, and Universal indicator paper.