ContactUsLinkComponent
Ward's® Introduction to Genetics: A Dihybrid Cross in Yeast Lab Activity
Ward's® Introduction to Genetics: A Dihybrid Cross in Yeast Lab Activity
Catalog # 470007-106
Supplier:  Avantor
undefined
Ward's® Introduction to Genetics: A Dihybrid Cross in Yeast Lab Activity
Catalog # 470007-106
Supplier:  Avantor
Specifications
  • Description:
    Dihybrid Cross in Yeast Lab Activity
  • No. of lab groups:
    6
  • Cat. No.:
    470007-106
Specifications
About this item
Mendel’s Law of Independent Assortment made easy.
Dihybrid crosses can be complex and abstract in nature, but this activity allows students to perform a cross that clearly demonstrates the properties of dominant and recessive alleles, genotypic and phenotypic ratios, independent assortment, and F1 and F2 inheritance patterns on just two Petri plates.
Before starting the experiment, students will predict the results of the cross. Next, they will perform the cross using simple microbiological techniques, and then analyze the results. You will get a teacher’s guide, student copymaster, and enough materials for six setups.
Kit Includes:
Sterile media and petri dishes required for performing crosses
Teacher's guide and student copymaster
Coupon redeemable for 8 different strains of yeast
Ordering information: This item contains a copupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Delivery: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.
Caution: Note: While the organism in this activity is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.