ContactUsLinkComponent
Ward's® Introduction to Accident Investigation Kit
Ward's® Introduction to Accident Investigation Kit
Catalog # 470175-744
Supplier:  Avantor
undefined
Ward's® Introduction to Accident Investigation Kit
Catalog # 470175-744
Supplier:  Avantor
Specifications
  • Description:
    WARD’S Introduction to Accident Investigation Kit
  • No. of lab groups:
    8
  • Cat. No.:
    470175-744
  • Supplier no.:
    WARD470175-744
Specifications
About this item
Address Concepts of Friction, Force and Energy
  • Reuseable Materials
  • Use of Physics to Solve Forensic Problems
Create a correlation between physics and forensics with activities that implement formulas and materials used by law enforcement. Students perform a series of experiments to learn how friction, force, and energy apply to interpreting skid marks discovered at a motor vehicle accident. Includes a teacher’s guide, student copymaster, and materials for eight setups.